Orders LinkContactUsLinkComponent
Raspberry Pi 4 Computers
USB-C power adapter
  470354-024
 :  03AH7034
Raspberry Pi 4 Computers
  470354-024
Raspberry Pi 4 Computers
Raspberry Pi 4 Computers
USB-C power adapter
$8.09
Each
Ground-breaking increases in speed and performance compared to the Raspberry Pi 3 Model B+.
  • Choose either 4 or 8 GB of RAM
  • Single board computer only; power adapter, accessories, display cable, and microSD card available separately
For the end user, Raspberry Pi 4 Model B provides desktop performance comparable to entry-level x86 PC systems. This product’s key features include a high-performance 64-bit quad-core processor, dual-display support at resolutions up to 4K via a pair of micro-HDMI ports, hardware video decode at up to 4Kp60, 4 GB of RAM, dual-band 2.4/5.0 GHz wireless LAN, Bluetooth 5.0, Gigabit Ethernet, USB 3.0, and PoE capability (via a separate PoE HAT add-on). The dual-band wireless LAN and Bluetooth have modular compliance certification, allowing the board to be designed into end products with significantly reduced compliance testing, improving both cost and time to market.
  • Description:
    USB-C power adapter