Orders LinkContactUsLinkComponent
Ziath DP5 Express Rack Reader
Ziath DP5 Express Rack Reader
Cyclops 1D linear code scanner for express
Catalog # 77321-928
Supplier:   AZENTA US, INC
Supplier #:  DP5-CYS-E
Ziath DP5 Express Rack Reader
Catalog # 77321-928
Supplier:  AZENTA US, INC
Ziath DP5 Express Rack Reader
Ziath DP5 Express Rack Reader
Cyclops 1D linear code scanner for express
Price
$958.35
Log in for availability
Each
Product Details & Documents
The express rack reader is a 2D datamatrix tube camera reader with an image capture time of only one second. Compatible with all 2D barcoded tube racks in SBS format, the express is the smallest rack scanner currently available and ideal for robotic integration.
  • Smallest footprint 2D barcode rack scanner
  • Fast camera-based scanner
  • Ideal for integration into large automated systems
  • Capable of scanning all SBS Rack formats
  • Patented technology allows imaging under difficult lighting conditions
  • Easy to set up and easy to use; free DP5 software upgrades for life
  • DP5 Express 2D scanner offers easy, out-of-the-box set up and is delivered pre-calibrated and ready to scan
  • Software includes remote control capability for integration applications
This instrument can scan all racks in the SBS format, regardless of the configuration or number of tubes, and is supplied with all the features needed for robotic integration.
Our new automatic rack type detection feature means that you no longer have to configure the Express for all rack types. Simply place your rack on the scanning window and the software will determine rack type and run the scan - easy.
Compatible with 24, 48, 96, 240, 384 position SBS racks. Compatible with all commercially available datamatrix 2D coded tubes. Export data as CSV, Text, XLS, XML, JSON, Python, DB, PNG or E-mail.
Operating system on host PC: Windows® 10, 11
Data output connector: USB3 or RJ45
With Cryoprotection: Prevents the scanner window from misting when scanning a cold rack from a freeze. The passive technology does not use heat or blown air.
 : CE marked.
 : Two-year warranty.
 : One reader system per case.
Specifications
  • Description:
    Cyclops 1D linear code scanner for express