BUFFER BETAGLUCAN
Catalog # MSPP-BG600
Supplier: CHARLES RIVER ENDOSAFE MS
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
Ethanol ≥70% (v/v), Biotechnology Grade
Supplier: VWR
VWR® Disposable Petri Dishes
Supplier: VWR International
CiDehol® ST Sterile 70% IPA with WFI, Decon Labs
Supplier: Decon Labs
VWR® Weighing Paper
Supplier: VWR International
Masterflex® I/P® Spooled Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
VWR®, Agarose I™
Supplier: VWR
Direct Thermal Cryotags, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences