Orders LinkContactUsLinkComponent
Erlenmeyer Flasks with Joint
Erlenmeyer Flasks with Joint
500 ml, 24/40
Catalog # 470353-640
Supplier #:  UNJTERFL500
Erlenmeyer Flasks with Joint
Catalog # 470353-640
Erlenmeyer Flasks with Joint
Erlenmeyer Flasks with Joint
500 ml, 24/40
Price
$47.61
New Customer Promotional Price
Log in for availability
Pack of 2
Product Details & Documents
Erlenmeyer flasks that can use a glass stopper.
  • Made from ASTM E-438 Type-1 Class A, Borosilicate 3.3 glass
  • Flasks include ground joints manufactured per ASTM E-1404, Type 2, Class-1 requirement
  • Flasks are printed in white color enamel
Borosilicate glass flasks are designed to provide the best combination of thermal shock resistance and physical strength. They feature 24/40 jointed sockets and graduations in durable white enamel.
Specifications
  • [ST] joint no.:
    24/40
  • Capacity:
    500 ml