Erlenmeyer Flasks with Joint
300 ml, 24/40
Catalog # 470353-644
Supplier: United Scientific Supplies
Supplier #: UNJTERFL300
Jump to:
Product Details & Documents
Erlenmeyer flasks that can use a glass stopper.
- Made from ASTM E-438 Type-1 Class A, Borosilicate 3.3 glass
- Flasks include ground joints manufactured per ASTM E-1404, Type 2, Class-1 requirement
- Flasks are printed in white color enamel
Borosilicate glass flasks are designed to provide the best combination of thermal shock resistance and physical strength. They feature 24/40 jointed sockets and graduations in durable white enamel.
Specifications
- [ST] joint no.:24/40
- Capacity:300 ml