LB AGAR PLATES WITH CARBENICILLIN-100 AN
Catalog # 89024-755
Supplier: Teknova
LB AGAR PLATES WITH CARBENICILLIN-100 AN
Catalog # 89024-755
Supplier: Teknova
Supplier Number: L1801
Frequently Bought Together
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
Vi-CELL BLU Reagent Kits, Beckman Coulter
Supplier: Beckman Coulter
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Officemate Assorted Magnets
Supplier: Janitorial Supplies
Paraformaldehyde 4% in PBS ready to use fixative reagent
Supplier: Biotium
LB Agar Plates, Teknova
Supplier: Teknova