HOLDER F/1-6MM ID SEPERATION COLUMN
Catalog # MSPP-GCH01S
Supplier: IMTAKT USA MS
HOLDER F/1-6MM ID SEPERATION COLUMN
Catalog # MSPP-GCH01S
Supplier: IMTAKT USA MS
Supplier Number: GCH01S
Frequently Bought Together
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
VWR® Weighing Paper
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International