Order Entry
United States
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine dihydrochloride 99%

3,3',5,5'-Tetramethylbenzidine dihydrochloride 99%

Catalog # 77275-822
Irritant
Supplier:   Ambeed
Supplier #:  A234741-1G
3,3',5,5'-Tetramethylbenzidine dihydrochloride 99%
Catalog # 77275-822
Supplier:  Ambeed
CAS Number:  64285-73-0
3,3',5,5'-Tetramethylbenzidine dihydrochloride 99%
CAS Number:  64285-73-0
Price
$18.00
Log in for availability
Each (1g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Caution: for Research Use Only

Synonyms:  Dihydrochloride TMB, TMB-HCl, TMB 2HCl, TMB dihydrochloride
  • CAS Number:64285-73-0
  • Formula:C₁₆H₂₀N₂·2HCl
  • MDL Number:MFCD00012961
  • Molecular Weight:313.27 g/mol

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.