Orders LinkContactUsLinkComponent
D-(-)-Glutamic acid 98%
Glass bottle, 100 g
  77275-148
 :   Ambeed
 :  A210714-100G
D-(-)-Glutamic acid 98%
  77275-148
 :  Ambeed
 :  6893-26-1
D-(-)-Glutamic acid 98%
Glass bottle, 100 g
 :  6893-26-1
$35.00
Each (100g)
 : for Research Use Only
 :  (R)-2-Aminopentanedioic acid, (R)-(-)-Glutamic acid, (R)-(-)-2-Aminopentanedioic acid, D(-)-2-Aminopentanedioic acid, D(-)-α-Glutamic acid, D(-)-1-Aminopropane-1,3-dicarboxylic acid, H-D-Glu-OH, (R)-(-)-α-Glutamic acid, (R)-(-)-1-Aminopropane-1,3-dicarboxylic acid
  •  :6893-26-1
  •  :C₅H₉NO₄
  •  :MFCD00063112
  •  :147.13 g/mol
  •  :333.8 °C (760 mm Hg)
  •  :200…202 °C
  •  :155.7 °C
  •  :Ambient
  • Pack type:
    Glass bottle
  • Size:
    100 g