TISSUE ARRAY PANCREAS CARCINOMA HUMAN
Catalog # MSPP-PA1002B
Supplier: TISSUEARRAY.COM LLC MS
TISSUE ARRAY PANCREAS CARCINOMA HUMAN
Catalog # MSPP-PA1002B
Supplier: TISSUEARRAY.COM LLC MS
Supplier Number: PA1002B
Frequently Bought Together
EDVOTEK Dual Gel Electrophoresis Apparatus
Supplier: Edvotek
Transfer Membranes, NuSep
Supplier: NuSep
Magnesium chloride hexahydrate for biochemistry
Supplier: G-Biosciences
Wax Paper Sheets, Cytiva
Supplier: Cytiva
BUFFER TRICINE 0.5M PH 7.5
Supplier: BIOWORLD MS
Accessories for Axygen® Horizontal Gel Box, 15 cm, Corning
Supplier: Corning
PROTEIN SIZE MARKER WIDE VEIW PRESTAINED
Supplier: FUJIFILM IRVINE SCIENTIFIC MS
Using CRISPR to treat Cystic Fibrosis
Supplier: Edvotek
DEFIBRINATED BLOOD SHEEP 100ML
Supplier: LAMPIRE BIOLOGICAL LAB MS
Reliant Precast DNA Minigels
Supplier: Lonza
BUFFER DNA/RNA HT 100ML
Supplier: Zymo Research
MOUSE SERUM US ORIGIN FOR COGLTN STUDIES
Supplier: Innovative Research Inc
ZYMOPURE BINDING BUFFER (110ML)
Supplier: Zymo Research
ARRAY TISSUE PDAC HUMAN 5UM TH 24CS
Supplier: TISSUEARRAY.COM LLC MS
Bovine Plasma with Citrate, Equitech- Bio, Inc.
Supplier: Equitech-Bio
Canine Plasma, Equitech- Bio, Inc.
Supplier: Equitech-Bio
SSC 20X Buffer, MP Biomedicals
Supplier: MP Biomedicals
Rat Plasma, Equitech- Bio
Supplier: Equitech-Bio