BUFFER STABILIZING REAGENT B 500ML
Catalog # MSPP-9100002
Supplier: CHEMOMETEC INC MS
BUFFER STABILIZING REAGENT B 500ML
Catalog # MSPP-9100002
Supplier: CHEMOMETEC INC MS
Supplier Number: 9100002
Frequently Bought Together
High Vacuum Grease, DuPont
Supplier: Liveo
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Sleeve Stoppers
Supplier: VWR International
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
Orion™ pH Buffer Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
EZ STANDARD PACK 1 PK8
Supplier: 0000014126
VIA1-CASSETTE FOR NUCLEOCOUNTER PK100
Supplier: CHEMOMETEC INC MS
BUFFER LYSIS REAGENT A100 500ML
Supplier: CHEMOMETEC INC MS
SYBR® Safe DNA Stain, 10,000 X, Edvotek
Supplier: Edvotek