COLUMN CADMIUM FOR NO3 DETERMINATION
Catalog # MSPP-50237A
Supplier: HACH COMPANY MS
COLUMN CADMIUM FOR NO3 DETERMINATION
Catalog # MSPP-50237A
Supplier: HACH COMPANY MS
Supplier Number: 50237A
Frequently Bought Together
Accessories for Barnstead GenPure xCAD Water Purification Systems, Thermo Scientific
Supplier: Thermo Fisher Scientific
Screw Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Sleeve Stoppers
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Vacuum Filtration Systems, Standard Line
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Sodium chloride 5 M, sterile-filtered
Supplier: Teknova
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
EDTA disodium salt dihydrate 99.0-101.0% ACS, VWR Chemicals BDH®
Supplier: VWR International
Plasmid+® Media, Thomson Instrument Company
Supplier: THOMSON INSTRUMENT CO
Pierce™ Reacti-Bind™ 96-Well Polystyrene Plates, Thermo Scientific
Supplier: Invitrogen
CARTRIDGE FLUORESCENCE MODULE 12-230 KDA
Supplier: 0000014126
Magnesium sulfate 1 M in aqueous solution
Supplier: G-Biosciences
Sodium chloride 0.9% in water normal saline solution
Supplier: G-Biosciences
COLUMN LC HYDRO-RP 80A 4UM 150X3MM
Supplier: 0000013469