TISSUE ARRAY STOMACH ADENOCARCINOMA HU
Catalog # MSPP-BS01012E
Supplier: TISSUEARRAY.COM LLC MS
TISSUE ARRAY STOMACH ADENOCARCINOMA HU
Catalog # MSPP-BS01012E
Supplier: TISSUEARRAY.COM LLC MS
Supplier Number: BS01012E
Frequently Bought Together
Buffer P3 (Yellow), Zymo Research
Supplier: Zymo Research
Protein Reducing Agents, G-Biosciences
Supplier: G-Biosciences
IPG Buffers, Cytiva
Supplier: Cytiva
Advansta WesternBright™ Sirius HRP Substrate Kits
Supplier: Advansta
10X Tris-Acetate-EDTA (TAE)
Supplier: Rockland Immunochemical
ARRAY TISSUE PDAC HUMAN 5UM TH 24CS
Supplier: TISSUEARRAY.COM LLC MS
Rat IL-1 beta ELISA Kit, Rockland Immunochemicals
Supplier: Rockland Immunochemical
PI MARKER 9.50 200UL
Supplier: PROTEIN SIMPLE MS
ELISA KIT 2 3 CYCLIC GAMPS
Supplier: 0000018898
Laemmli SDS sample buffer, non-reducing (4X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
PI MARKER ICE280/ICE3 7.05 200UL
Supplier: PROTEIN SIMPLE MS
Sodium dodecyl sulfate (SDS) solution 10% (w/v), Molecular Biology Grade
Supplier: Promega Corporation
Electrophoresis Running Buffers, G-Biosciences
Supplier: G-Biosciences
Protease Inhibitor Mix, Cytiva
Supplier: Cytiva
ScanLater™ Western Blot Assay Kit, Molecular Devices
Supplier: Molecular Devices
N,N,N',N'-Tetramethylethylenediamine (TEMED) for electrophoresis
Supplier: Thermo Scientific Chemicals
Lysis Buffer 7X (Blue), Zymo Research
Supplier: Zymo Research
1X PBS Buffer with 1% Casein
Supplier: Rockland Immunochemical
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
GelBond® Film
Supplier: Lonza