Orders LinkContactUsLinkComponent
Eisco Plastic Scoops
Eisco Plastic Scoops
Plastic scoops, 5 ml
Catalog # 470343-957
Supplier:   Wards
Supplier #:  CH0461BPK12
Eisco Plastic Scoops
Catalog # 470343-957
Supplier:  Wards
Eisco Plastic Scoops
Eisco Plastic Scoops
Plastic scoops, 5 ml
Price
$17.75
Log in for availability
Pack of 12
Product Details & Documents
Pack of 12 or 6 weighing scoops with labeled handles and flat bottoms.
  • Made of heat- and chemical-resistant polypropylene plastic
  • Scoop handles are labeled with capacity for easy identification
  • Flat bottom design allows scoops to rest upright for weighing and filling
Autoclavable and dishwasher safe for easy cleaning.
Specifications
  • Capacity:
    5 ml
  • Description:
    Plastic scoops