TISSUE ARRAY COLORECTAL CANCER 150CASES
Catalog # MSPP-CRM1502
Supplier: TISSUEARRAY.COM LLC MS
TISSUE ARRAY COLORECTAL CANCER 150CASES
Catalog # MSPP-CRM1502
Supplier: TISSUEARRAY.COM LLC MS
Supplier Number: CRM1502
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
Corning® Internal Thread Cryogenic Vials, Corning
Supplier: Corning
VWR® Traceable® Timer with Auto Shutoff
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International