ASSAY DEVELOP CUSTOM CELL-BASED CALCIUM
Catalog # 77147-910
Supplier: MULTISPAN INC
Related Information
MULTISCREEN Custom Potency Cell-based Assay Development Services
Application Notes
Multispan MULTISCREEN™ Products and Services
Product Literature
Frequently Bought Together