ContactUsLinkComponent
1-(6-Amino-3,5-difluoropyridin-2-yl)-8-chloro-6-fluoro-7-(3-hydroxyazetidin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, Purity: 98%, CAS Number: 189279-58-1, Appearance: Form: solid, Storage: Inert atmosphere, Room Temperature, Size: 50mg
Catalog # 77109-742
Supplier:  Ambeed
1-(6-Amino-3,5-difluoropyridin-2-yl)-8-chloro-6-fluoro-7-(3-hydroxyazetidin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, Purity: 98%, CAS Number: 189279-58-1, Appearance: Form: solid, Storage: Inert atmosphere, Room Temperature, Size: 50mg
Catalog # 77109-742
Supplier:  Ambeed
Supplier Number:  A139239-50MG