Ecom Seating Polyurethane Tractor Stools
Stool with chrome foot ring and chrome base finish, Blue, Stationary glides, 23 - 33½"
Catalog # 470352-440
Supplier: E COM SEATING SE
Supplier #: PTHBST-CR-CF-RG-BLU
Jump to:
Product Details & Documents
Attractive design featuring tractor style seat and ergonomic adjustments for seat height and seat tilt.
- Pneumatic seat height adjustment
- Rugged polyurethane, easily cleaned, chemical-resistant seat
- Available with stationary glides or casters
Ordering information: 15 year parts and lifetime pneumatic lift warranty.
Delivery information: Ships unassembled.
Specifications
- Color:Blue
- Description:Stool with chrome foot ring and chrome base finish
- Seat height:23 - 33½"
- Foot style:Stationary glides