Ecom Seating Polyurethane Tractor Stools
Stool, no foot ring, black fiberglass base finish, Black, Stationary glides, 16 - 21"
Catalog # 470352-392
Supplier: E COM SEATING SE
: PTDHSL-RG-RG-BLK
Jump to:
Product Details & Documents
Attractive design featuring tractor style seat and ergonomic adjustments for seat height and seat tilt.
- Pneumatic seat height adjustment
- Rugged polyurethane, easily cleaned, chemical-resistant seat
- Available with stationary glides or casters
: 15 year parts and lifetime pneumatic lift warranty.
: Ships unassembled.
Specifications
- Color:Black
- Description:Stool, no foot ring, black fiberglass base finish
- Seat height:16 - 21"
- Foot style:Stationary glides