BUFFER B MSD GOLD READ 90 ML
MESO-R60AM-2
Frequently Bought Together
VWR® Broken Glass Disposal Cartons
: VWR International
VWR® Disposable Serological Pipettes
: VWR International
96-Well Black and White Microplates, Polystyrene, Greiner Bio-One
: Greiner Bio-One
VWR® Disposable Petri Dishes
: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
: Sakura Finetek
VWR® Universal Pipette Tips
: VWR International
VWR® Nitrile Examination Gloves, Black
: VWR International
WASH BUFFER 20X WITH SURFACTANT 100ML
: Meso Scale Diagnostics
HIL-6 RA QKIT SIXPAK
: R&D Systems
SteriPettes, Kimble Chase, DWK Life Sciences
: DWK Life Sciences (KIMBLE)
BUFFER B READ MSD GOLD 200ML
: Meso Scale Diagnostics
KIT HIGH-CAPACITY RNA-TO-CDNA
: Invitrogen
VWR® Chemical-Resistant Laboratory Marker
: VWR International
ANTIBODY RPLEX HUMAN ER F21AKP-3
: Meso Scale Diagnostics
DILUENT 57 50ML
: Meso Scale Diagnostics