MAGicBead™ cfDNA Isolation Kit
Product Details & Documents
Unique, magnetic bead surface chemistry that facilitates unparalleled cfDNA recovery from biofluids.
- Highest yield: Robust cfDNA isolation from up to 10 ml plasma with unique MAGicBead™ surface technology
The MAGicBead™ cfDNA isolation kit features a unique, magnetic bead surface chemistry that facilitates unparalleled cfDNA recovery from biofluids. Input volumes can range up to 10 ml of plasma. It is compatible with all open automation platforms and a wide range of sample types (serum, saliva, urine, etc.). This kit features a simple, streamlined extraction process with minimal number of steps and does not require the bead drying step common with other similar procedures. Eluted cfDNA is free of enzymatic inhibitors that can interfere with PCR or other sensitive downstream applications, including Next-Gen sequencing. It is validated on all sensitive downstream assays, such as NGS and RT-qPCR.
Specifications
Specifications
- Description:MAGicBead™ cfDNA elution buffer
- Includes:Elution buffer designed to work only with the MAGicBead™s™ chemistry
- Cat. No.:77020-290
- Prep size:Scalable
- Refine target molecule:cfDNA
- Target molecule:DNA
- Method / Format:Magnetic beads
- Sample type:Biofluids (serum, plasma, saliva)



