Orders LinkContactUsLinkComponent
Quick-Lock Connectors
Quick-Lock Connectors
Quick-lock connector set II, 1 capillary tube and 1 safety cap
Catalog # 76948-966
Supplier:   SCAT AMERICAS, INC.
Supplier #:  360189
Quick-Lock Connectors
Catalog # 76948-966
Quick-Lock Connectors
Quick-Lock Connectors
Quick-lock connector set II, 1 capillary tube and 1 safety cap
Price
$119.76
Log in for availability
Pack of 1
Minimum order quantity is 1
Product Details & Documents
Easy and safe connection of capillary tubing and SCAT safety caps using quick couplings.
  • Made of chemically resistant glass-filled polypropylene, PEEK® materials
  • Leak protection thanks to integrated shut-off valves
  • Closed system for drip-free disconnection with very low air entrapment
  • Minimizes contamination and no need to purge air bubbles from the system
  • Suitable for capillaries with outer diameter 1.6, 2.3, and 3.2 mm
  • User-friendly, ergonomic design, enables safe connection
  • Up to vacuum 15 psi, 1,0 bar pressure resistance
Connect and disconnect with a 'click', without dripping or air bubbles. The valve closes automatically when disconnected. Thanks to the included PFA fittings, all common hose diameters can be connected.
The quick-lock connector sets are suitable for connecting 2 capillaries (set 1) or one capillary and one SCAT safety cap (set 2).
Specifications
  • Description:
    Quick-lock connector set II
  • For use with:
    1 capillary tube and 1 safety cap
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Quick-lock connector (f), includes 3× PFA-fitting (1,6/2,3/3,2 mm Int.Ø)
    Quick-lock connector (f), includes 3× PFA-fitting (1,6/2,3/3,2 mm Int.Ø)
    76948-960
    Log in for availability
    Each - $58.94
Product Information
  • Catalog #
    Availability
    Price
  • Quick-lock connector (m), includes 3× PTFE-ferrule (1,6/2,3/3,2 mm Int.Ø)
    Quick-lock connector (m), includes 3× PTFE-ferrule (1,6/2,3/3,2 mm Int.Ø)
    76948-962
    Log in for availability
    Each - $74.66
Product Information
  • Catalog #
    Availability
    Price
  • Quick-lock connector (m), includes 3× PFA-fitting (1,6/2,3/3,2 mm Int.Ø)
    Quick-lock connector (m), includes 3× PFA-fitting (1,6/2,3/3,2 mm Int.Ø)
    76948-968
    Log in for availability
    Each - $58.94