N-BOC-N-BIS PEG2-ACID 97% 437.5 500MG
Catalog # MSPP-BP2341050
Supplier: BROADPHARM MS
N-BOC-N-BIS PEG2-ACID 97% 437.5 500MG
Catalog # MSPP-BP2341050
Supplier: BROADPHARM MS
Supplier Number: BP-23410-500MG
Frequently Bought Together
VWR® Media/Storage Bottles with GL Screw Caps
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
2-Propanol ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
VWR® Weighing Paper
Supplier: VWR International
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International