DRYING TRAP MEDIUM 50-100ML ACETONITRILE
Catalog # MSPP-DMT-1975
Supplier: CHEMGENES MS
DRYING TRAP MEDIUM 50-100ML ACETONITRILE
Catalog # MSPP-DMT-1975
Supplier: CHEMGENES MS
Supplier Number: DMT-1975
Frequently Bought Together
ROCK SALT
Supplier: INDUSTRIAL SUPPORT INC.
Ammonia solution 28.0 - 30.0% (NH₃ basis) in water ACS, Fluka™
Supplier: Honeywell Research Chemicals
Laboratory Warm/Incubation Rooms, Precision Controlled Environments, Darwin Chambers Company
Supplier: Darwin Chambers Company
2-Acetylpyridine 98%
Supplier: Ambeed
MKM100 MTIM-1/KIM-1 QKIT (1 KT)
Supplier: R&D Systems
Ward's® Introduction to Agarose Gel Electrophoresis Lab Activity
Supplier: Avantor
SOLUTION BOND EPITOPE RETRIEVAL 1L
Supplier: Leica Biosystems MS
Oakton® pH Buffer Solutions, Cole-Parmer
Supplier: OAKTON INSTRUMENTS MS
Human Recombinant PSAP/Prosaposin Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
MastAssure™ Bacterial Agglutinating Antisera
Supplier: Hardy Diagnostics
Fluid A, Hardy Diagnostics
Supplier: Hardy Diagnostics
Omega Supreme Cleanroom HEPA Vac, Atrix
Supplier: Atrix International
INDICATOR CHEM STEAM PK=BX250
Supplier: Steris
Potassium chloride
Supplier: G-Biosciences
KWIK-STIK <i>Bacillus pumilus</i> ATCC® 700814™, Microbiologics
Supplier: MICROBIOLOGICS, INC.
DRYING TRAP XL UPTO 4L ACETONITRILE PK10
Supplier: 0000031866
FUSING AGENT HP 3D400 500 ML
Supplier: CIMQUEST, INC MS
TRAPS DRYING SMALL 10-50ML PACK 10
Supplier: CHEMGENES MS
Fluid Thioglycollate Medium, Dehydrated, HiMedia Laboratories
Supplier: HiMedia
IGM ELISA KIT HUMAN PLASMA
Supplier: Novus Biologicals