STANDARD ENDOTHALL 100UG/ML IN MEOH 1ML
Catalog # MSPP-P-183S
Supplier: ACCUSTANDARD INC MS
STANDARD ENDOTHALL 100UG/ML IN MEOH 1ML
Catalog # MSPP-P-183S
Supplier: ACCUSTANDARD INC MS
Supplier Number: P-183S
Frequently Bought Together
Nalgene® Marking Pens, Thermo Scientific
Supplier: Thermo Fisher Scientific
VWR® Silicone Laboratory Benchtop Mat
Supplier: VWR International
EDVOTEK Dual Gel Electrophoresis Apparatus
Supplier: Edvotek
Reusable Plastic Animal Cages, Tecniplast®
Supplier: Tecniplast Usa
Bytac® Teflon® Resin Surface Protectors, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
SpotDrops™ Nitrate Reagent B, Naphthylamine, Hardy Diagnostics
Supplier: Hardy Diagnostics
Caffeic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
PAD CORRUGATED 11 X 14 150LB
Supplier: 0000014096
Emodin ≥96.0% (by HPLC)
Supplier: TCI America
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
PI MARKER ICE280/ICE3 5.12 200UL
Supplier: 0000014126
Caffeic acid, Sigma-Aldrich®
Supplier: MilliporeSigma
Potato Dextrose Agar, Hardy Diagnostics
Supplier: Hardy Diagnostics
VECTOR 8E+12VG 2E+13VG/ML CONC 4X0.1ML
Supplier: VIROVEK, INC. MS
Chrysophanol ≥96.0% (by HPLC)
Supplier: TCI America
Static Rat/Hamster Cages, 2154F
Supplier: Tecniplast Usa