CRYOVIAL 1ML INT THREAD SS STER CS500
Catalog # MSPP-229913B
Supplier: CELLTREAT SCIENTIFIC PRODUCTS MS
CRYOVIAL 1ML INT THREAD SS STER CS500
Catalog # MSPP-229913B
Supplier: CELLTREAT SCIENTIFIC PRODUCTS MS
Supplier Number: 229913B
Frequently Bought Together
WHEATON® CryoELITE® Cryogenic Vials, Freestanding, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Thermo Scientific™ Matrix™ Screw Top Tube Caps, Cap Trays and Cap Inserts
Supplier: Thermo Fisher Scientific
Non-Coded Sample Storage Tubes, 0.5 ml, 96-format, External Thread, Azenta Life Sciences
Supplier: AZENTA US, INC
BioX Cryogenic Vials, Barcoded
Supplier: TWD Scientific
CRYOVIAL INT THREAD SS STER 1.8ML CS100
Supplier: VistaLab
Thermo Scientific™ Matrix™ Screw Top Tubes in Barcoded Latch Racks, 0.5 ml
Supplier: Thermo Fisher Scientific
CRYOVIAL 1ML EXT THREAD SS STER CS100
Supplier: VistaLab
Cryo.s™ Cryogenic Storage Vials, Polypropylene, Sterile, Greiner Bio-One
Supplier: Greiner Bio-One
CryoClear™ Cryogenic Vials, Internal Threads, Globe Scientific
Supplier: Globe Scientific
CryoPure Tubes with External Thread
Supplier: SARSTEDT INC
CryoELITE® Cryogenic Vials, Freestanding, External Thread, Shelf Pack, Wheaton
Supplier: DWK Life Sciences
Tri-Coded Sample Storage Tubes, 0.65 ml, 24-format, External Thread, Azenta Life Sciences
Supplier: AZENTA US, INC
CryoPure Tubes with Internal Thread and Silicone Seal
Supplier: SARSTEDT INC
Pacific Hemostasis® Fibrinogen Assay Kit, Thermo Scientific
Supplier: Thermo Scientific
Thermo Scientific™ Racks for Matrix™ 2D Barcoded Storage Tubes
Supplier: Thermo Fisher Scientific
TruCool™ Hinged Lid CryoBoxes
Supplier: AZENTA US, INC