WELL DEEP BLOCK FORMAT JCSG PLUST HT 1ML
Catalog # MSPP-HR2150
Supplier: HAMPTON RESEARCH MS
WELL DEEP BLOCK FORMAT JCSG PLUST HT 1ML
Catalog # MSPP-HR2150
Supplier: HAMPTON RESEARCH MS
Supplier Number: HR2-150
Frequently Bought Together
Biotube™ Cluster Tube System, Simport
Supplier: Simport Scientific
Cell Culture Dishes, quadriPERM®
Supplier: SARSTEDT INC
Micrewtube® Microcentrifuge Tube with Screw Cap, Simport Scientific
Supplier: Simport Scientific
Axygen® Microcentrifuge Screw Caps, Corning
Supplier: Corning
Micrewtube™ Microcentrifuge Tubes, Simport Scientific
Supplier: Simport Scientific
Microcentrifuge Screw Caps Closures for Micrewtubes™ Centrifuge Tubes, Simport
Supplier: Simport Scientific
Watson Brand Microtube Accessories, Watson Bio Lab
Supplier: Watson Bio Lab
Accessories for CoolBox™ XT/2XT, Corning®
Supplier: Corning
SPOOL PIPETTE MOSQUITO TIPS 9 MM PITCH
Supplier: SPT LABTECH LTD MS
SOLUTION DDM/CHS PRE-MADE 10:1 50ML
Supplier: ANATRACE MS
Screwcap Tubes without Caps, Watson Bio Lab
Supplier: Watson Bio Lab
PETRI DISH W GRIP 100MM X 15MM S CS300
Supplier: VistaLab
Pump Valves and Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
G-Tube® Microcentrifuge Tubes, 0.6 ml, Bio Plas
Supplier: Bio Plas
Storage and Assay Microplates
Supplier: Agilent Technologies
CRYSTAL SCREEN MCSG-1 1.7 ML BLOCK
Supplier: ANATRACE MS
Microcentrifuge Tubes, Caplugs
Supplier: Caplugs