SCREW MICRO TUBE ONLY 2.0ML SS NS CS1000
Catalog # MSPP-230831
Supplier: CELLTREAT SCIENTIFIC PRODUCTS MS
SCREW MICRO TUBE ONLY 2.0ML SS NS CS1000
Catalog # MSPP-230831
Supplier: CELLTREAT SCIENTIFIC PRODUCTS MS
Supplier Number: 230831
Frequently Bought Together
Screw Cap Tubes, 5.0 ml, MTC Bio
Supplier: GeneMate
VWR® Microcentrifuge Tubes for Boiling Applications
Supplier: VWR International
VWR® Disposable Microcentrifuge Tubes
Supplier: VWR International
VWR® Microcentrifuge Tubes with Socket Screw-Cap
Supplier: VWR International
LoBind Protein or Genomic Microcentrifuge Tubes, Eppendorf
Supplier: Eppendorf
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
VWR® Microcentrifuge Tubes, Low-Binding
Supplier: VWR International
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: Eppendorf
Locking Microcentrifuge Tubes, Globe Scientific
Supplier: Globe Scientific
VWR® Micro Centrifuge Tube with Flat Screw-Cap
Supplier: VWR International
Eppendorf® Flex-Tubes® Microtubes, 1.5 ml
Supplier: Eppendorf
VWR® High Performance Microcentrifuge Tubes, PP
Supplier: VWR International
Zobell Marine Agar and Broth, HiMedia Laboratories
Supplier: HiMedia
TUBE PROTEIN LOBIND PCRCLEAN 1.5ML PK500
Supplier: EPPENDORF NORTH AMERICA MS