Order Entry
United States
Orders LinkContactUsLinkComponent
 

Dapsone 98%

Glass bottle, 100 g
  76874-400
 
 :   Ambeed
 :  A240646-100G
Dapsone 98%
  76874-400
 :  Ambeed
 :  80-08-0
Dapsone 98%

Glass bottle, 100 g

 :  80-08-0
 
$21.00
   
Each (100g)
 
   
 

 : for Research Use Only

 :  4-Aminophenyl sulfone, 1-Amino-4-((4-aminophenyl)sulfonyl)benzene, 4,4'-sulfonyldianiline, 4,4'-Sulfonyldianiline, 4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Diamino diphenyl sulfone, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone, 4-[(4-Aminophenyl)sulfonyl]aniline
  •  :80-08-0
  •  :C₁₂H₁₂N₂O₂S
  •  :MFCD00007887
  •  :248.31 g/mol
 

 

  • Pack type:
    Glass bottle
  • Size:
    100 g

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.