BUFFER TISSUE LYSIS TRUXTRAC PROTIN 25ML
Catalog # MSPP-520284
Supplier: COVARIS INC MS
BUFFER TISSUE LYSIS TRUXTRAC PROTIN 25ML
Catalog # MSPP-520284
Supplier: COVARIS INC MS
Supplier Number: 520284
Frequently Bought Together
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
VWR® Sterile 70% Isopropanol
Supplier: VWR International
Precision pH Test Strips
Supplier: Precision Laboratories
CAPS 8 AFA-TUBE TPX STRIP PK12
Supplier: COVARIS INC MS