CYTOSOFT T-75 FLASKS 0.5 KPA
Catalog # 104019-148
Supplier: ADVANCED BIOMATRIX, INC. MS
CYTOSOFT T-75 FLASKS 0.5 KPA
Catalog # 104019-148
Supplier: ADVANCED BIOMATRIX, INC. MS
Supplier Number: 5324-6EA
Frequently Bought Together
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
CYTOSOFT T-75 FLASKS 16 KPA
Supplier: ADVANCED BIOMATRIX, INC. MS