ACCU-SCOPE® C-Mount Microscope Camera Adapters
1× C-mount adapter for 1" chip sensors
Catalog # 470351-132
Supplier: ACCU-SCOPE INC SE
Supplier #: 00-2010-1
Jump to:
Product Details & Documents
Microscope camera adapters are designed to fit ACCU-SCOPE's EXC-400, EXC-350, 3001-LED or 3001 microscopes.
- Multiple sizes available
Adapter can be focused to match the focus of the digital image to the image seen through the eyepieces.
Specifications
- Description:1× C-mount adapter for 1" chip sensors