Orders LinkContactUsLinkComponent
micro:bit v2
micro:bit V2 Club Bundle (V2 GO Pk 10)
Catalog # 470350-732
Supplier:   PARALLAX INC.
 :  32719
micro:bit v2
Catalog # 470350-732
Supplier:  PARALLAX INC.
micro:bit v2
micro:bit v2
micro:bit V2 Club Bundle (V2 GO Pk 10)
Price
$262.69
Log in for availability
Pack of 10
Product Details & Documents
These ARE the micro:bits you've been looking for!
  • Write programs in your choice of languages: blockly, python
The micro:bit v2 module is a compact programmable board with built in LED display, buttons, tilt sensor, speaker, and radio, made for friendly computer education. The BBC micro:bit v2 will be completely familiar to anyone who has used the original device. It’s the same size, shape and looks very similar — and functions the same way. Programs written for the prior version run on version 2 with the same editors. However, the micro:bit v2.0 features several significant additions related to program memory (3.5× the code space for cyber:bot Python scripts), a touch-pad button, speaker and microphone. These hardware additions expand the kinds of applications students may build.
Some additional refinements include a notched edge connector, power LED, and an exposed, gold-plated antenna to identify the radio/Bluetooth component.
 : Module includes (1) micro:bit controller. Go Bundle includes (1) BBC Micro:bit controllers, (1) USB cable, (1) battery holder, (2) AAA batteries, user guide. Club Bundle includes (10) Go Bundles.
Specifications
  • Description:
    micro:bit V2 Club Bundle (V2 GO Pk 10)