Orders LinkContactUsLinkComponent
cyber:bot Robots
cyber:bot Robots
cyber:bot robot
Catalog # 470350-736
Supplier:   PARALLAX INC.
Supplier #:  32700
cyber:bot Robots
Catalog # 470350-736
Supplier:  PARALLAX INC.
cyber:bot Robots
cyber:bot Robots
cyber:bot robot
Price
$288.29
Log in for availability
Each
Product Details & Documents
All the power and robustness of a Parallax rover, but with the accessibility of a micro:bit.
  • Python-programmable, powered by the popular BBC micro:bit
  • Beginner-friendly, and no special tools needed
  • Build DIY sensor circuits on the breadboard that use touch, visible light, and infrared light
  • Free online Robotics and Cybersecurity tutorials and micro:bit robotics curriculum resources for educators
The cyber:bot robot puts the BBC micro:bit module on the Parallax small robot! The cyber:bot robot adds a tangible hardware dimension to Python-focused computer science, CTE and cybersecurity programs. This form factor is a proven STEM success in classrooms from middle school into college, leveraging the add-ons available for this chassis. A pre-programmed onboard Propeller Multicore Microcontroller assists the micro:bit, handling the real-time servo motor control and sensors built on the breadboard. This enables more robust robotic applications that go beyond what the micro:bit can do on its own.
Delivery information: Kit includes (1) micro:bit module, (1) cyber:bot board, (1) small robot chassis, (2) parallax continuous rotation servos, (2) small robot wheel and tire, (1) small robot electronics pack, (1) small robot hardware pack, (1) 5AA batteryholder, barrel jack, (1) USB A to micro B cable, (1) extra hardware for cyber:bot board, parallax pocket screwdriver.
Specifications
  • Description:
    cyber:bot robot