ACETONITRILE HPLC GRADE LIQUID AMBER 4L
Catalog # MSPP-EW-88092
Supplier: Cole Parmer
ACETONITRILE HPLC GRADE LIQUID AMBER 4L
Catalog # MSPP-EW-88092
Supplier: Cole Parmer
Supplier Number: EW-88000-92
Frequently Bought Together
VWR® Sleeve Stoppers
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
CiDehol® ST Sterile 70% IPA with WFI, Decon Labs
Supplier: Decon Labs
Sodium chloride 5 M, sterile-filtered
Supplier: Teknova
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
ROCK SALT
Supplier: INDUSTRIAL SUPPORT INC.
Acetonitrile ≥99.9% (by GC), BAKER ANALYZED® HPLC, Ultra Gradient for HPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
VWR® Talon® Lab Frame Feet
Supplier: VWR International
Water, AquaPur™ ST USP, purified, sterile
Supplier: Decon Labs
Tryptic Soy Agar (TSA), Soybean Casein Digest Agar
Supplier: Hardy Diagnostics
BioBall® SingleShot 30
Supplier: bioMerieux
Firefly Gene Transformation Kit
Supplier: Avantor
Ward's® Introduction to Agarose Gel Electrophoresis Lab Activity
Supplier: Avantor
Low Form Zirconium Crucibles, Thermo Scientific
Supplier: Thermo Scientific Chemicals
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
Hexagel™ Electrophoresis Chamber, Edvotek
Supplier: Edvotek
DxH 500 Hematology Analyzer, Beckman
Supplier: Beckman Coulter
SYBR® Safe DNA Stain, 10,000 X, Edvotek
Supplier: Edvotek