FLOW RESTRICTOR TUBING 0.005IN ID 0.3 ML
Catalog # MSPP-1100-0161
Supplier: Pickering Labs
FLOW RESTRICTOR TUBING 0.005IN ID 0.3 ML
Catalog # MSPP-1100-0161
Supplier: Pickering Labs
Supplier Number: 1100-0161
Frequently Bought Together
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Universal Pipette Tips
Supplier: VWR International
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
CARBAMATE HYDROLYSIS REAGENT 950ML CS4
Supplier: Pickering Labs