ARTIFICIAL SALIVA NOT STABILIZED 200 ML
Catalog # MSPP-1700-0304
Supplier: Pickering Labs
ARTIFICIAL SALIVA NOT STABILIZED 200 ML
Catalog # MSPP-1700-0304
Supplier: Pickering Labs
Supplier Number: 1700-0304
Frequently Bought Together
VWR® Weighing Paper
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Razor Blades
Supplier: VWR International
2-Propanol ≥99.0% USP, VWR Chemicals BDH®
Supplier: BDH
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Disposable Petri Dishes
Supplier: VWR International
Simulated gastric fluid (without pepsin) (0.2% NaCl) (w/v) in hydrochloric acid 0.7%
Supplier: Ricca Chemical