Order Entry
United States
Orders LinkContactUsLinkComponent
Vector PCX Derivatization Instruments

Vector PCX Derivatization Instruments

Vector, dual pump, 0.5 ml, 120 V (streptomycin)
Catalog # MSPP-1154-4001
Marketsource
Supplier:   Pickering Labs
Supplier #:  1154-4001
Vector PCX Derivatization Instruments
Catalog # MSPP-1154-4001
Supplier:  Pickering Labs
Marketsource
Vector PCX Derivatization Instruments

Vector, dual pump, 0.5 ml, 120 V (streptomycin)

Price
$34,830.77
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Vector PCX provides the selectivity and sensitivity required for most standard post-column applications while being reliable and easy to use. Since Vector PCX does not have a column oven it is important to use the HPLC column oven to ensure stable column temperature and prevent retention time drifts and separation problems.

  • Independently adjustable, low-pulsation inert pumps
  • PEEK liquid manifolds, Integrated gas manifold
  • Easy access and monitoring of fluidics
  • Simple control interface (no computer required)
  • Reduced bench space profile
  • Sapphire pistons with automatic piston wash

Dimensions: 43×21.6×41.2 cm (17×8.75×16 inches). Weight: 12.5 kg (27.6 lbs) – Duplex. Electrical: 100 to 120 V, 50/60 Hz 1.7 A, 200 W or 200 to 240 V, 50/60 Hz, 0.8 A, 200 W. Mains voltage ±10% of nominal. Installation (over voltage) category II, pollution degree 2. Environmental: altitude up to 6500 ft. Ambient temperature 15 to 25 ºC. Relative humidity up to 80% at 31 ºC. Adjustable from 0.05 to 2.00 ml/minute against back-pressures of up to 2000 psi.

One liter capacity (2 L reservoirs available). Maintained under inert gas pressure to inhibit oxidation of oxygensensitive reagents. Valve built into reservoir cap permits sparging during reagent preparation. Reagent reservoirs fitted with 3.1 mm (¹/₈") Ext.Ø, oxygen-impermeable Air Barrier tubing for oxygen-sensitive reagents and/or with 3.1 mm (¹/₈") Ext.Ø FEP tubing.

Specifications
  • Description:
    Vector, dual pump, 0.5 ml, 120 V (streptomycin)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.