VESICLE ECOLI OUTER MEMBRANE 100UG
Catalog # MSPP-TLRL-OMV
Supplier: Invivogen
VESICLE ECOLI OUTER MEMBRANE 100UG
Catalog # MSPP-TLRL-OMV
Supplier: Invivogen
Supplier Number: TLRL-OMV-1
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
Corning® Centrifuge Tubes, 15 ml
Supplier: Corning
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Multiwell Cell Culture Plates
Supplier: VWR International
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
Corning® Internal Thread Cryogenic Vials, Corning
Supplier: Corning
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Anti-CD44 Rat Monoclonal Antibody (Biotin) [clone: IM7]
Supplier: Biolegend
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International