NORMOCIN PREVENTING MYCOPLASMA 500 MG
Catalog # MSPP-ANT-NR-1
Supplier: Invivogen
NORMOCIN PREVENTING MYCOPLASMA 500 MG
Catalog # MSPP-ANT-NR-1
Supplier: Invivogen
Supplier Number: ANT-NR-1
Frequently Bought Together
VWR® Signature™ Lab Coats made with DuPont™ Tyvek® IsoClean® Material
Supplier: VWR International
Precellys® CK14 Lysing Kit, Soft Tissue Homogenizing, Bertin
Supplier: BERTIN CORP
HEK-Blue™ Selection, InvivoGen
Supplier: Invivogen
Blasticidin S hydrochloride solution 10 mg/ml in HEPES ≥95% (by HPLC)
Supplier: Invivogen
Anti-NCOA4 Mouse Monoclonal Antibody [clone: 439CT10.4.4]
Supplier: Abgent
CELL LINE JURKAT FL GREEN 3X10-6CELLS
Supplier: CELLS ONLINE, LLC MS
Anti-SDF-1alpha (CXCL12) Rabbit Polyclonal Antibody (Biotin)
Supplier: Peprotech
ASSAY UPLEX HUMAN IP10 K151UFK-4
Supplier: Meso Scale Diagnostics
Normocin™ - Antimicrobial Reagent, InvivoGen
Supplier: Invivogen
MEDIUM ALKALINEPHOSPHATASE DETECTION 5ML
Supplier: Invivogen
Human Recombinant SCF (from <i>E. coli</i>)
Supplier: Peprotech
RECOM IL-23 >95% BY SDS-PAGE HUMAN 10UG
Supplier: Novus Biologicals
CDS AGONIST VACV-70 LYOVEC COMPLEX 100UG
Supplier: Invivogen
Blasticidin, Invivogen
Supplier: Invivogen
Hydrochloric acid R1 250 g/L
Supplier: Ricca Chemical
Monarch® Genomic DNA Purification Kits, New England Biolabs
Supplier: New England Biolabs (NEB)
Mouse Recombinant IL-22 (from <i>E. coli</i>)
Supplier: R&D Systems
ZEOCIN 1G CS10
Supplier: Invivogen
QUANTI BLUE SOLUTION 100X LIQUID 10ML
Supplier: Invivogen