TISSUE CULTR MULTPL WELL PLATE PK5
Catalog # MSPP-229197
Supplier: CELLTREAT SCIENTIFIC PRODUCTS MS
TISSUE CULTR MULTPL WELL PLATE PK5
Catalog # MSPP-229197
Supplier: CELLTREAT SCIENTIFIC PRODUCTS MS
Supplier Number: 229197
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
Trypsin Protease, HyClone™
Supplier: Cytiva
VWR® LTS Compatible Pipette Tips
Supplier: VWR International
VWR® Ultra High Performance Centrifuge Tubes
Supplier: VWR International
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
96-Well Microplates, Polystyrene, Clear, Greiner Bio-One
Supplier: Greiner Bio-One
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® PCR Strip Tubes & Caps 8-Strip
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Cell Culture Plates Treated for Increased Cell Attachment, Sterile
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International
GripTip Pipette Tips, Racked, Integra Biosciences
Supplier: INTEGRA MS
Nuclease P1
Supplier: New England Biolabs (NEB)
MASTER MIX 2 RAPID TAQ 5X1ML
Supplier: VAZYME INTERNATIONAL LLC MS
ASSAY LUMINESCENT CELL VIABILITY 100ML
Supplier: VAZYME INTERNATIONAL LLC MS
Thiophosphoryl chloride ≥98%
Supplier: Strem Chemicals Inc