Orders LinkContactUsLinkComponent
Dropping Bottle
Dropping Bottle
7 ml, LDPE
Catalog # 470349-808
Supplier #:  PDB007PK12
Dropping Bottle
Catalog # 470349-808
Dropping Bottle
Dropping Bottle
7 ml, LDPE
Price
$5.20
Log in for availability
Pack of 12
Product Details & Documents
Dispense solutions accurately, one drop at a time.
  • Ideal for storing and dispensing small volumes
  • Translucent design
  • LDPE with leakproof PP cap
  • BPA-free
Dropping bottles are useful for adding small amounts of a liquid. With a small squeeze the amount of liquid being dispensed can be controlled. Manufactured from translucent and low density polyethylene. Caps are made from polypropylene. Assembled bottles are sold as packs.
Specifications
  • Capacity:
    7 ml
  • Material:
    LDPE