Orders LinkContactUsLinkComponent
Slide Storage Boxes, Hinged, Plastic
Slide Storage Boxes, Hinged, Plastic
Slide storage box, hinged, Plastic, 100 slides
Catalog # 470349-780
Supplier #:  62213
Slide Storage Boxes, Hinged, Plastic
Catalog # 470349-780
Slide Storage Boxes, Hinged, Plastic
Slide Storage Boxes, Hinged, Plastic
Slide storage box, hinged, Plastic, 100 slides
Price
$10.30
Log in for availability
Each
Product Details & Documents
Store slides securely and safely.
  • Engraved numbers
  • Secure closing clasp
  • Key card for easy reference
Sturdy two-piece molded slide boxes hold standard 3×1" microscope slides. The hinged top has a reference card for labeling the corresponding slide description. The numbers on the opposite side are engraved for easy reference. These boxes are made from high impact polystyrene.
Specifications
  • Capacity:
    100 slides
  • Description:
    Slide storage box, hinged
  • Material:
    Plastic