Slide Storage Boxes, Hinged, Plastic
Slide storage box, hinged, Plastic, 50 slides
Catalog # 470349-770
Supplier: United Scientific Supplies
Supplier #: 62212
Jump to:
Product Details & Documents
Store slides securely and safely.
- Engraved numbers
- Secure closing clasp
- Key card for easy reference
Sturdy two-piece molded slide boxes hold standard 3×1" microscope slides. The hinged top has a reference card for labeling the corresponding slide description. The numbers on the opposite side are engraved for easy reference. These boxes are made from high impact polystyrene.
Specifications
- Capacity:50 slides
- Description:Slide storage box, hinged
- Material:Plastic