Orders LinkContactUsLinkComponent
Genomic DNA Kits
Genomic DNA Kits
Genomic DNA separation gel, 240 ml, For use with the Fragment Analyzer systems and the DNF-467, DNF-468, and DNF-488 kits
Catalog # 76645-104
Supplier #:  DNF-270-0240
Genomic DNA Kits
Catalog # 76645-104
Genomic DNA Kits
Genomic DNA Kits
Genomic DNA separation gel, 240 ml, For use with the Fragment Analyzer systems and the DNF-467, DNF-468, and DNF-488 kits
Price
$1,070.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Genomic DNA quality control of whole genome libraries, including sizing of gDNA extractions.
  • Broad concentration range - The HS gDNA 50 kb kit has a concentration range of 0.3 to 12 ng/µl, while the gDNA 50 kb kit covers 25 to 250 ng/µl
  • High and standard sensitivity kits - Minimize sample dilutions and choose the kit that has the right input concentration range for your needs
  • Reliable Sizing - Sizing range from 75 bp to 60 kb ensures accurate and precise sizing of gDNA samples
  • Low sample volume - Only 1 to 2 µl of sample is required, minimizing sample loss to QC steps
The genomic DNA kits are available for the automated assessment of gDNA sizing and integrity on the fragment analyzer systems. The kits are designed for quality control of gDNA extraction samples used in whole genome sequencing, metagenomics, and large structural variant analysis. They are also suited for the analysis of degraded DNA, such as formalin-fixed parafilm embedded (FFPE) samples.
Sample preparation is simplified by eliminating dilution steps with separate genomic DNA kits for high (HS) and low concentrated gDNA samples. A broad sizing range allows researchers to accurately size gDNA samples through 60 kb.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
  • Description:
    Genomic DNA separation gel, 240 ml
  • Details:
    For use with the Fragment Analyzer systems and the DNF-467, DNF-468, and DNF-488 kits