Bioanalyzer High Sensitivity Protein Analysis
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
High sensitivity electrophoretic separation of proteins and peptides up to 250 kD.
- A ready-to-use system featuring standardized assays and pre-packaged reagent kits which are suitable to analyze samples up to 250 kD
- After sample labeling, fast results are achieved with automated electrophoresis of 10 samples in 30 minutes
- Highly sensitive with an accuracy down to silver stain levels, approximately 1 pg/µl of labeled protein on chip
- Fully operational across a large linear dynamic range with four orders of magnitude
- Highly precise impurity detection, down to 0.05% and lower
Perform high sensitivity protein electrophoresis with the 2100 bioanalyzer system. The high sensitivity protein 250 assay provides a broad dynamic range with sensitivity down to silver stain comparable levels. Efficiency is enhanced by eliminating handling of SDS-PAGE slab gels, staining or, imaging steps.
The system provides a fast, automated, objective and flexible solution for protein and peptide characterization, quality control, and impurity detection in the low picogram range. Compatible sample types include protein lysates, purified protein and peptides, antibodies, and protein stability tests.
Caution: For research use only. Not for use in diagnostic procedures.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Agilent high sensitivity protein 250 labeling kit
- Details:Reagents for labeling reaction only, sufficient for 100 samples