Orders LinkContactUsLinkComponent
ADC® e-Sphyg 3+
ADC® e-Sphyg 3+
Wall mount
Catalog # 470349-512
Supplier #:  9003M
ADC® e-Sphyg 3+
Catalog # 470349-512
ADC® e-Sphyg 3+
ADC® e-Sphyg 3+
Wall mount
Price
$324.90
Log in for availability
Each Case of 2
Product Details & Documents
NIBP monitor with Adcuff+.
  • Easy to clean, bladderless, cuffs
  • Auto mode for quick readings in as little as 30 seconds
  • Manual mode for auscultation with a stethoscope
  • Customizable three-reading average mode with individual measurement display
  • Last-reading memory and irregular heartbeat detection
  • Three-year warranty on unit and cuffs
  • Self-standing design and built-in handle
ADC’s all-new e-sphyg 3 captures blood pressure measurements with the touch of a button. Designed for busy professionals who value both accuracy and affordability, the e-sphyg 3 is the feature-packed, clinical-grade solution you’ve been waiting for. The Adcuff+ model features our all-new, one-piece reusable nylon blood pressure cuffs.
Auto mode for quick readings in as little as 30 seconds. Manual mode for auscultation with a stethoscope. Customizable three-reading average mode with individual measurement display. Last-reading memory and irregular heartbeat detection. Oscillometric measurement with for systolic and diastolic blood pressures, pulse rate, and mean arterial pressure (MAP). High-capacity rechargeable battery and AC power adapter, standard. For use with adult and pediatric patients. Three-year warranty on unit. The e-sphyg 3 features a self-standing design and built-in handle for portability. Made without phthalates or natural rubber latex. Inspected and packaged in the USA.
Specifications
  • Description:
    Wall mount