Orders LinkContactUsLinkComponent
ADC® e-Sphyg 3+
ADC® e-Sphyg 3+
e-Sphyg 3+ monitor (small adult, adult and large adult)
Catalog # 470349-510
Supplier #:  9003K-MCC
ADC® e-Sphyg 3+
Catalog # 470349-510
ADC® e-Sphyg 3+
ADC® e-Sphyg 3+
e-Sphyg 3+ monitor (small adult, adult and large adult)
Price
$551.50
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
NIBP monitor with Adcuff+.
  • Easy to clean, bladderless, cuffs
  • Auto mode for quick readings in as little as 30 seconds
  • Manual mode for auscultation with a stethoscope
  • Customizable three-reading average mode with individual measurement display
  • Last-reading memory and irregular heartbeat detection
  • Three-year warranty on unit and cuffs
  • Self-standing design and built-in handle
ADC’s all-new e-sphyg 3 captures blood pressure measurements with the touch of a button. Designed for busy professionals who value both accuracy and affordability, the e-sphyg 3 is the feature-packed, clinical-grade solution you’ve been waiting for. The Adcuff+ model features our all-new, one-piece reusable nylon blood pressure cuffs.
Auto mode for quick readings in as little as 30 seconds. Manual mode for auscultation with a stethoscope. Customizable three-reading average mode with individual measurement display. Last-reading memory and irregular heartbeat detection. Oscillometric measurement with for systolic and diastolic blood pressures, pulse rate, and mean arterial pressure (MAP). High-capacity rechargeable battery and AC power adapter, standard. For use with adult and pediatric patients. Three-year warranty on unit. The e-sphyg 3 features a self-standing design and built-in handle for portability. Made without phthalates or natural rubber latex. Inspected and packaged in the USA.
Specifications
  • Description:
    e-Sphyg 3+ monitor (small adult, adult and large adult)