RACK FLIPPER 4 WAY W/TRAY ASSORTED
Catalog # MSPP-1148C96
Supplier: THOMAS SCIENTIFIC MS
RACK FLIPPER 4 WAY W/TRAY ASSORTED
Catalog # MSPP-1148C96
Supplier: THOMAS SCIENTIFIC MS
Supplier Number: 1148C96
Frequently Bought Together
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Corning® Internal Thread Cryogenic Vials, Corning
Supplier: Corning
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
Methanol ≥99.8% ACS, VWR Chemicals BDH®
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Parafilm® M Sealing Film
Supplier: Bemis Company, Inc.
Corning® PBS (Phosphate-Buffered Saline), Corning
Supplier: Corning
ALCH ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
VWR® Single-Edged Razor Blades
Supplier: VWR International