ALKALINE PHOSPHATASE DETECTN MEDIUM 20ML
Catalog # MSPP-REP-QBS3
Supplier: Invivogen
ALKALINE PHOSPHATASE DETECTN MEDIUM 20ML
Catalog # MSPP-REP-QBS3
Supplier: Invivogen
Supplier Number: REP-QBS3
Frequently Bought Together
VWR® Razor Blades
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
Dispensing Tips, EFD
Supplier: EFD INC.- CUSTOMER SERVICE TE
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
BD Microtainer® Tubes, BD Diagnostics
Supplier: BD
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
VWR® Chemical-Resistant Laboratory Marker
Supplier: VWR International
NORMOCIN PREVENTING MYCOPLASMA 500 MG
Supplier: Invivogen
WASH BUFFER 20X WITH SURFACTANT 100ML
Supplier: Meso Scale Diagnostics
HEK-Blue™ Selection, InvivoGen
Supplier: Invivogen
NORMOCIN ANTIMICROBIAL REAGENT 250MG
Supplier: Invivogen
Accessories for U-2D™ 96-Well Plate Sample Control System, MicroSolv
Supplier: MicroSolv Technology Corp